Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HSFY1 Full-length ORF (NP_149099.2, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00086614P01L

 View more versions of this product

Catalog No. 89-948-006


Add to cart

Description

Description

This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MAHVSSETQDVSPKDELTASEASTRSPLCEHTFPGDSDLRSMIEEHAFQVLSQGSLLESPSYTVCVSEPDKDDDFLSLNFPRKLWKIVESDQFKSISWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKIQQNFQRSAFLATFLSEEKESSVLSKLKFYYNPNFKRGYPQLLVRVKRRIGVKNASPISTLFNEDFNKKHFRAGANMENHNSALAAEASEESLFSASKNLNMPLTRESSVRQIIANSSVPIRSGFPPPSPSTSVGPSEQIATDQHAILNQLTTIHMHSHSTYMQARGHIVNFITTTTSQYHIISPLQNGYFGLTVEPSAVPTRYPLVSVNEAPYRNMLPAGNPWLQMPTIADRSAAPHSRLALQPSPLDKYHPNYN
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSF2L|HSFY
HSFY1
Yes
NP_149099.2
Liquid
86614
HSFY1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAHVSSETQDVSPKDELTASEASTRSPLCEHTFPGDSDLRSMIEEHAFQVLSQGSLLESPSYTVCVSEPDKDDDFLSLNFPRKLWKIVESDQFKSISWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKIQQNFQRSAFLATFLSEEKESSVLSKLKFYYNPNFKRGYPQLLVRVKRRIGVKNASPISTLFNEDFNKKHFRAGANMENHNSALAAEASEESLFSASKNLNMPLTRESSVRQIIANSSVPIRSGFPPPSPSTSVGPSEQIATDQHAILNQLTTIHMHSHSTYMQARGHIVNFITTTTSQYHIISPLQNGYFGLTVEPSAVPTRYPLVSVNEAPYRNMLPAGNPWLQMPTIADRSAAPHSRLALQPSPLDKYHPNYN
RUO
HSFY1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit