Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HSN2 Full-length ORF (NP_998820.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00378465P01S

 View more versions of this product

Catalog No. 89-948-017


Add to cart

Description

Description

This intronless gene is poorly expressed and lies within an intron of WNK1 gene (GeneID:65125) on chr 12. Mutations in this gene are associated with hereditary sensory and autonomic neuropathy, type II, an autosomal recessive disorder characterized by impairment of pain, temperature, and touch sensation owing to reduction or absence of peripheral sensory neurons. It is proposed that this gene product may play a role in the development and/or maintenance of peripheral sensory neurons or their supporting Schwann cells. [provided by RefSeq]

Sequence: MYELLVLFMLIQPQSMAHPCGGTPTYPESQIFFPTIHERPVSFSPPPTCPPKVAISQRRKSTSFLEAQTHHFQPLLRTVGQSLLPPGGSPTNWTPEAVVMLGTTASRVTGESCEIQVHPMFEPSQVYSDYRPGLVLPEEAHYFIPQEAVYVAGVHYQARVAEQYEGIPYNSSVLSSPMKQIPEQKPVQGGPTSSSVFEFPSGQAFLVGHLQNLRLDSGLGPGSPLSSISAPISTDATRLKFHPVFVPHSAPAVLTHNNESRSNCVFEFHVHTPSSSSGEGGGILPQRVYRNRQVAVDLNQEELPPQSVGLHGYLQPVTEEKHNYHAPELTVSVVEPIGQNWPIGSPEYSSDSSQITSSDPSDFQSPPPTGGAAAPFGSDVSMPFIHLPQTVLQESPLFFCFPQGTTSQQVLTASFSSGGSALHPQVIGKLPQLF
Specifications

Specifications

NP_998820.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSAN2
HSN2
Yes
wheat germ expression system
Liquid
378465
HSN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYELLVLFMLIQPQSMAHPCGGTPTYPESQIFFPTIHERPVSFSPPPTCPPKVAISQRRKSTSFLEAQTHHFQPLLRTVGQSLLPPGGSPTNWTPEAVVMLGTTASRVTGESCEIQVHPMFEPSQVYSDYRPGLVLPEEAHYFIPQEAVYVAGVHYQARVAEQYEGIPYNSSVLSSPMKQIPEQKPVQGGPTSSSVFEFPSGQAFLVGHLQNLRLDSGLGPGSPLSSISAPISTDATRLKFHPVFVPHSAPAVLTHNNESRSNCVFEFHVHTPSSSSGEGGGILPQRVYRNRQVAVDLNQEELPPQSVGLHGYLQPVTEEKHNYHAPELTVSVVEPIGQNWPIGSPEYSSDSSQITSSDPSDFQSPPPTGGAAAPFGSDVSMPFIHLPQTVLQESPLFFCFPQGTTSQQVLTASFSSGGSALHPQVIGKLPQLF
RUO
HSN2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit