Learn More
Novus Biologicals™ Human HSP40/DNAJB1 ELISA Kit (Colorimetric)
Human HSP40/DNAJB1 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276440
Catalog No. NBP276440
Add to cart
Description
Description
The Human HSP40/DNAJB1 ELISA Kit (Colorimetric) measures human HSP40/DNAJB1 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 to 10 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human HSP-40 in samples. No significant cross-reactivity or interference between Human HSP-40 and analogues was observed.
Human
1 Kit
3337
0.10 ng/mL
Storage is content dependent.
DNAJB1
Biological Fluids, Plasma, Serum