Learn More
Raybiotech IncSupplier Diversity Partner Human HSP70 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHHSP701
For cell culture supernatants, Heparin and/or EDTA treated plasma, and serum samples. Citrate is not recommended. Sensitivity: 2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair also can recognize mouse and rat HSP70
Catalog No. 50-194-9360