Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Human HSP90 alpha Platinum ELISA Kit Non-distribution product as customer accommodation.

For the quantitative detection of human HSP90 alpha.

Manufacturer:  Invitrogen™ BMS2090

 View more versions of this product

Catalog No. 50-112-8684


Add to cart

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human HSP90 alpha
Biotin-Conjugate anti-human HSP90 alpha monoclonal antibody
Streptavidin-HRP
Human HSP90 alpha Standard lyophilized, 20ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 (MIM 140572), a constitutive form. Other HSP90 proteins are found in endoplasmic reticulum (HSP90B1; MIM 191175) and mitochondria (TRAP1; MIM 606219) (Chen et al., 2005 [PubMed 16269234]).
Specifications

Specifications

P07900
Biotin
3320
Adhesive Plate Covers
96 tests
0.03 ng/mL
100μL (1:25 prediluted)
Human
1 hr 20 min
Human
Heat Shock Protein 90 alpha
8.6%
0.156-10 ng/mL
ELISA
Colorimetric Microplate Reader
ELISA Kit
RUO
Cell Lysate , Plasma, Serum
Cell Lysate,100 μL|Plasma,10 μL|Serum,10 μL
2-8°C
4 hr
HSP90AA1
10%
HRP
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit