Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HSPA1L Full-length ORF (AAH34483.1, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003305P01S

 View more versions of this product

Catalog No. 89-948-026


Add to cart

Description

Description

This gene encodes a 70kDa heat shock protein. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which also encode isoforms of the 70kDa heat shock protein. [provided by RefSeq]

Sequence: MATAKGIAIGIDLGTTYSCVGVFQHGKVEIIANDQGNRTTPSYVAFTDTERLIGDAAKNQVAMNPQNTVFDAKRLIGRKFNDPVVQADMKLWPFQVINEGGKPKVLVSYKGENKAFYPEEISSMVLTKLKETAEAFLGHPVTNAVITVPAYFNDSQRQATKDAGVIAGLNVLRIINEPTAAAIAYGLDKGGQGERHVLIFDLGGGTFDVSILTIDDGIFEVKATAGDTHLGGEDFDNRLVSHFVEEFKRKHKKDISQNKRAVRRLRTACERAKRTLSSSTQANLEIDSLYEGIDFYTSITRARFEELCADLFRGTLEPVEKALRDAKMDKAKIHDIVLVGGSTRIPKVQRLLQDYFNGRDLNKSINPDEAVAYGAAVQAAILMGDKSEKVQDLLLLDVAPLSLGLETAGGVMTALMKRNSTIPTKQTQIFTTYSDNQPGVLIQVYEGERAMTKDNNLLGRFDLTGIPPAPRGVPQIEVTFDIDANGILNVTAMDKSTGKVNKITIANDKGRLSKEEIERMVLDAEKYKAEDEVQREKIAAKNALESYAFNMKSVVSDEGLKGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVD
Specifications

Specifications

AAH34483.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
96.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSP70-1L|HSP70-HOM|HSP70T|hum70t
HSPA1L
Yes
wheat germ expression system
Liquid
3305
HSPA1L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATAKGIAIGIDLGTTYSCVGVFQHGKVEIIANDQGNRTTPSYVAFTDTERLIGDAAKNQVAMNPQNTVFDAKRLIGRKFNDPVVQADMKLWPFQVINEGGKPKVLVSYKGENKAFYPEEISSMVLTKLKETAEAFLGHPVTNAVITVPAYFNDSQRQATKDAGVIAGLNVLRIINEPTAAAIAYGLDKGGQGERHVLIFDLGGGTFDVSILTIDDGIFEVKATAGDTHLGGEDFDNRLVSHFVEEFKRKHKKDISQNKRAVRRLRTACERAKRTLSSSTQANLEIDSLYEGIDFYTSITRARFEELCADLFRGTLEPVEKALRDAKMDKAKIHDIVLVGGSTRIPKVQRLLQDYFNGRDLNKSINPDEAVAYGAAVQAAILMGDKSEKVQDLLLLDVAPLSLGLETAGGVMTALMKRNSTIPTKQTQIFTTYSDNQPGVLIQVYEGERAMTKDNNLLGRFDLTGIPPAPRGVPQIEVTFDIDANGILNVTAMDKSTGKVNKITIANDKGRLSKEEIERMVLDAEKYKAEDEVQREKIAAKNALESYAFNMKSVVSDEGLKGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVD
RUO
HSPA1L
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit