Learn More
Abnova Human HYAL1 Partial ORF (NP_009296, 60 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$384.10 - $599.28
Specifications
Name | HYAL1 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 36.74kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-948-147 |
Abnova
H00003373Q01L |
25 ug |
Each for $599.28
|
|
89-948-146 |
Abnova
H00003373Q01S |
10 ug |
Each for $384.10
|
|
Description
This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: ANPGQTFRGPDMTIFYSSQLGTYPYYTPTGEPVFGGLPQNASLIAHLARTFQDILAAIPAPDFSGLAVIDWEAWRPRWAFNWDTKDIYRQRSRALVQAQHSpecifications
HYAL1 (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
36.74kDa |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.