Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HYLS1 Full-length ORF (NP_659451.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00219844P01

Catalog No. 89-948-163


Add to cart

Description

Description

This gene encodes a protein localized to the cytoplasm. Mutations in this gene are associated with hydrolethalus syndrome. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MEELLPDGQIWANMDPEERMLAAATAFTHICAGQGEGDVRREAQSIQYDPYSKASVAPGKRPALPVQLQYPHVESNVPSETVSEASQRLRKPVMKRKVLRRKPDGEVLVTDESIISESESGTENDQDLWDLRQRLMNVQFQEDKESSFDVSQKFNLPHEYQGISQDQLICSLQREGMGSPAYEQDLIVASRPKSFILPKLDQLSRNRGKTDRVARYFEYKRDWDSIRLPGEDHRKELRWGVREQMLCRAEPQSKPQHIYVPNNYLVPTEKKRSALRWGVRCDLANGVIPRKLPFPLSPS
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.8kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32915|HLS
HYLS1
Yes
NP_659451.1
Liquid
219844
HYLS1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEELLPDGQIWANMDPEERMLAAATAFTHICAGQGEGDVRREAQSIQYDPYSKASVAPGKRPALPVQLQYPHVESNVPSETVSEASQRLRKPVMKRKVLRRKPDGEVLVTDESIISESESGTENDQDLWDLRQRLMNVQFQEDKESSFDVSQKFNLPHEYQGISQDQLICSLQREGMGSPAYEQDLIVASRPKSFILPKLDQLSRNRGKTDRVARYFEYKRDWDSIRLPGEDHRKELRWGVREQMLCRAEPQSKPQHIYVPNNYLVPTEKKRSALRWGVRCDLANGVIPRKLPFPLSPS
RUO
HYLS1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit