Learn More
Raybiotech IncSupplier Diversity Partner Human ICAM-2/CD102 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHICAM21
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.17 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human ICAM-2.
Catalog No. 50-194-9365