Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Human ICAM-2/CD102 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191181
Catalog No. NBP191181
Description
Description
The Human ICAM-2/CD102 ELISA Kit (Colorimetric) measures human ICAM-2/CD102 in cell culture supernatant, serum, plasma (edta, citrate, heparin), amniotic fluid, urine, whole blood.
Specifications
Specifications
0.75 - 24.0 U/mL
3384
ELISA Kit
0.30 U/mL
Human (Hu)
ICAM-2/CD102
ICAM2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sICAM-2, Biotin-Conjugate anti-human sICAM-2 monoclonal antibody, Streptavidin-HRP, Human sICAM-2 Standard lyophilized, 24.00U/mL upon reconstitution, Streptavidin-HRP Diluent, Standard Buffer Diluent (10x) Biotin Conjugate Diluent, Wash Buffer Concentrate 200x, Substrate Solution (tetramethyl-benzidine and buffered hydrogen peroxide), Stop Solution (1.8N H2SO4), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine, whole blood
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Immunology
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2