Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ICAM1 (05362, D1-D2) Partial Recombinant Protein fused with the Fc of the immunoglobulin superfamily with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P6178

Catalog No. 89-948-188


Add to cart

Description

Description

This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor. [provided by RefSeq]

Specifications

Specifications

05362
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
23kDa
Escherichia coli expression system
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
BB2|CD54|P3.58
ICAM1
Escherichia coli
His
Escherichia coli expression system
Liquid
3383
ICAM1 (D1-D2) (Human) Recombinant Protein
100 ug
RUO
ICAM1
Human ;Human
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit