Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ICMT Full-length ORF (NP_036537.1, 1 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023463P01

Catalog No. 89-948-205


Add to cart

Description

Description

This gene encodes the third of three enzymes that posttranslationally modify isoprenylated C-terminal cysteine residues in certain proteins and target those proteins to the cell membrane. This enzyme localizes to the endoplasmic reticulum. Alternative splicing may result in other transcript variants, but the biological validity of those transcripts has not been determined. [provided by RefSeq]

Sequence: MAGCAARAPPGSEARLSLATFLLGASVLALPLLTRAGLQGRTGLALYVAGLNALLLLLYRPPRYQIAIRACFLGFVFGCGTLLSFSQSSWSHFGWYMCSLSLFHYSEYLVTAVNNPKSLSLDSFLLNHSLEYTVAALSSWLEFTLENIFWPELKQITWLSVTGLLMVVFGECLRKAAMFTAGSNFNHVVQNEKSDTHTLVTSGVYAWFRHPSYVGWFYWSIGTQVMLCNPICGVSYALTVWRFFRDRTEEEEISLIHFFGEEYLEYKKRVPTGLPFIKGVKVDL
Specifications

Specifications

NP_036537.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.3kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSTE14|MGC39955|MST098|MSTP098|PCCMT|PCMT|PPMT
ICMT
Yes
wheat germ expression system
Liquid
23463
ICMT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGCAARAPPGSEARLSLATFLLGASVLALPLLTRAGLQGRTGLALYVAGLNALLLLLYRPPRYQIAIRACFLGFVFGCGTLLSFSQSSWSHFGWYMCSLSLFHYSEYLVTAVNNPKSLSLDSFLLNHSLEYTVAALSSWLEFTLENIFWPELKQITWLSVTGLLMVVFGECLRKAAMFTAGSNFNHVVQNEKSDTHTLVTSGVYAWFRHPSYVGWFYWSIGTQVMLCNPICGVSYALTVWRFFRDRTEEEEISLIHFFGEEYLEYKKRVPTGLPFIKGVKVDL
RUO
ICMT
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit