Learn More
Human IFN-alpha Multi-Subtype ELISA Kit, R&D Systems
formulated to detect 14 out of 15 identified human IFN-α subtypes
Manufacturer:  R&D Systems 411051
 View more versions of this product
Catalog No. 411051
  Check Availability
Add to cart
Includes:
  • Pre-coated microtiter plate(s)
  • Plate sealers
  • Wash Solution Concentrate
  • Human Interferon Alpha Standard, 10,000 pg/ml
  • Dilution Buffer
  • Antibody Concentrate
  • HRP Conjugate Concentrate
  • Con
Specifications
Description
VeriKine™ Human IFN-α Multi-Subtype ELISA kit is specifically formulated to detect 14 out of 15 identified human IFN-α subtypes. They are: IFN-αA, IFN-α2, IFN-αD, IFN-αB2, IFN-αC, IFN-αG, IFN-αH, IFN-αI, IFN-αJ1, IFN-αK, IFN-α1, IFN-α4a, IFN-α4b, and IFN-αWA.
Description & Specifications
Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3440
1 Plate
12.5pg/mL
Cell Culture Supernates (100μL)
Human IFN-α
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates
Human
Sufficient materials to run ELISAs on at least one 96-well plate