Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IFN-alpha Multi-Subtype ELISA Kit
Human IFN-alpha Multi-Subtype ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: R&D Systems™ 411052
Includes:
- Pre-coated microtiter plate(s)
- Plate sealers
- Wash Solution Concentrate
- Human Interferon Alpha Standard, 10,000 pg/ml
- Dilution Buffer
- Antibody Concentrate
- HRP Conjugate Concentrate
- Con
Description
The Human IFN-alpha Multi-Subtype ELISA Kit quantifies human IFN-alpha in cell culture supernates (100 ul).Specifications
12.50 to 500 pg/mL (Cell Culture Supernates), 156.00 to 5000 pg/mL (Cell Culture Supernates) | |
Biotin | |
Solid Phase Sandwich ELISA | |
Human IFN-alpha | |
Human | |
3440 | |
Cell Culture Supernates (100 μL) | |
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D |
12.5pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Cell Culture Supernatant | |
Sufficient materials to run ELISAs on at least five 96-well plates | |
ELISA | |
5 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction