Learn More
Novus Biologicals™ Human IFN-gamma ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191174
Catalog No. NBP191174
Add to cart
Description
Description
The Human IFN-gamma ELISA Kit (Colorimetric) measures human IFN-gamma in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
1.6 - 100pg/mL
3458
ELISA Kit
0.99pg/mL
Human (Hu)
IFN-gamma
IFNG
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-gamma, Biotin-Conjugate anti-human IFN-gamma monoclonal antibody, Streptavidin-HRP, Human IFN-gamma Standard lyophilized, 200ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma