Learn More
Novus Biologicals™ Human IFN-omega/IFNW1 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191178
Catalog No. NBP191178
Add to cart
Description
Description
The Human IFN-omega/IFNW1 ELISA Kit (Colorimetric) measures human IFN-omega/IFNW1 in cell culture supernatant, serum, plasma (edta, citrate, heparin).
Specifications
Specifications
4.7 - 300pg/mL
3467
ELISA Kit
1.5pg/mL
Human (Hu)
IFN-omega/IFNW1
IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IFN-omega, Biotin-Conjugate anti-human IFN-omega monoclonal antibody, Streptavidin-HRP, Human IFN-omega Standard, 6ng/mL, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Storage of components varies. See protocol for specific instructions.
IFNW1