Learn More
Abnova™ Human IFNA2 (P01563, 166 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4105
Catalog No. 89-948-346
Add to cart
Specifications
Specifications
P01563
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
19.4kDa
Escherichia coli expression system
100 ug
RUO
IFNA|INFA2|MGC125764|MGC125765
IFNA2
Escherichia coli
None
Escherichia coli expression system
Lyophilized
3440
IFNA2 (Human) Recombinant Protein
≥95% by SDS-PAGE and HPLC
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
IFNA2
The specific activity as determined in a viral resistance assay using VSV-WISH cells was found to be greater than 1 x 108 IU/mg.
Yes