Learn More
Abnova™ Human IFNA2 (P01563, 171 a.a.) Full-length Recombinant Protein with C-terminal His tag
Used for Func, SDS-PAGE, WB-Re
Manufacturer:  Abnova™ P5916
Catalog No. 89-948-348
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Sequence: CDLPQTHSLGSRRTLMLLAQMRKISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPLMKEDSILAVRKYFQRITLYLKEKKYSPCAWEVVRAEIMRSFSLSTNLQESLRSKEHHHHHH
Specifications
Specifications
P01563
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis, Western Blot (Recombinant protein)
Lyophilized
20.1kDa
Non-transgenic plants (Nicotiana benthamiana) expression system
>97% by SDS-PAGE
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.04 EU/μg protein
IFNA2
The specific activity was determined by a quantitative gene reported bioassay using human Type I interferon-sensitive. The activity is < 8 x 108 units/mg in reference to a viral resistance assay using bovine kidney MDBK cells.
Plants
His
Non-transgenic plants (Nicotiana benthamiana) expression system
Lyophilized
3440
IFNA2 (Human) Recombinant Protein
Sequential chromatography (FPLC)
0.3 ug/lane in 15% SDS-polyacrylamide gel and stained with Coomassie blue
CDLPQTHSLGSRRTLMLLAQMRKISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPLMKEDSILAVRKYFQRITLYLKEKKYSPCAWEVVRAEIMRSFSLSTNLQESLRSKEHHHHHH
RUO
IFNA|INFA2|MGC125764|MGC125765
IFNA2
Human ;Human
Yes