Learn More
Abnova™ Human IFNA2 (P01563, 24 a.a. - 188 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3622
Catalog No. 89-948-345
Add to cart
Description
Description
Sequence: CDLPQTHSLGSRRTLMLLAQMRKISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPLMKEDSILAVRKYFQRITLYLKEKKYSPCAWEVVRAEIMRSFSLSTNLQESLRSKE
Specifications
Specifications
P01563
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
19kDa
Escherichia coli expression system
>90% by SDS-PAGE and HPLC
CDLPQTHSLGSRRTLMLLAQMRKISLFSCLKDRHDFGFPQEEFGNQFQKAETIPVLHEMIQQIFNLFSTKDSSAAWDETLLDKFYTELYQQLNDLEACVIQGVGVTETPLMKEDSILAVRKYFQRITLYLKEKKYSPCAWEVVRAEIMRSFSLSTNLQESLRSKE
RUO
IFNA|INFA2|MGC125764|MGC125765
IFNA2
Escherichia coli
None
Escherichia coli expression system
Lyophilized
3440
IFNA2 (Human) Recombinant Protein
Ion exchange column and HPLC reverse phase column
100 ug
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg (1 EU/μg)
IFNA2
The ED50 was determined by viral resistance assay and was found to be not less than 5 x 108 IU/mg.
Yes