Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human IFNA2 (Q86UP4, 165 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P4106
Specifications
Q86UP4 | |
Lyophilized | |
19.3kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
IFNA2 | |
The specific activity as determined in a viral resistance assay using bovine kidney MDBK cells was found to be 2.6 x 108 units/mg. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and HPLC |
Functional Study, SDS-PAGE | |
3440 | |
IFNA2 (Human) Recombinant Protein | |
100 ug | |
RUO | |
IFNA/INFA2/MGC125764/MGC125765 | |
IFNA2 | |
E. coli | |
None | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction