Learn More
Abnova™ Human IFNG (P01579, 24 a.a. - 166 a.a.) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3624
Catalog No. 89-948-392
Add to cart
Description
Description
Interferon-gamma (IFNG), or type II interferon, is a cytokine critical for innate and adaptive immunity against viral and intracellular bacterial infections and for tumor control. Aberrant IFNG expression is associated with a number of autoinflammatory and autoimmune diseases. The importance of IFNG in the immune system stems in part from its ability to inhibit viral replication directly, but most importantly derives from its immunostimulatory and immunomodulatory effects. IFNG is produced predominantly by natural killer (NK) and natural killer T (NKT) cells as part of the innate immune response, and by CD4 (MIM 186940) and CD8 (see MIM 186910) cytotoxic T lymphocyte (CTL) effector T cells once antigen-specific immunity develops (Schoenborn and Wilson, 2007 [PubMed 17981204]).[supplied by OMIM]
Sequence: QDPYVKEAENLKKYFNAGHSDVADNGTLFLGILKNWKEESDRKIMQSQIVSFYFKLFKNFKDDQSIQKSVETIKEDMNVKFFNSNKKKRDDFEKLTNYSVTDLNVQRKAIHELIQVMAELSPAAKTGKRKRSQMLFRGRRASQ
Specifications
Specifications
P01579
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
17kDa
Escherichia coli expression system
>90% by SDS-PAGE and HPLC
QDPYVKEAENLKKYFNAGHSDVADNGTLFLGILKNWKEESDRKIMQSQIVSFYFKLFKNFKDDQSIQKSVETIKEDMNVKFFNSNKKKRDDFEKLTNYSVTDLNVQRKAIHELIQVMAELSPAAKTGKRKRSQMLFRGRRASQ
RUO
IFG|IFI
IFNG
Escherichia coli
None
Escherichia coli expression system
Lyophilized
3458
IFNG (Human) Recombinant Protein
Ion exchange column and HPLC reverse phase column
100 ug
Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg (1 EU/μg)
IFNG
The ED50 was determined by a viral resistance assay of mouse L929 cells and was found to be ≤1ng/mL, corresponding to a specific activity of ≥ 1 x 106 units/mg.
Yes