Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IFNK Partial ORF (NP_064509, 35 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00056832Q01S

 View more versions of this product

Catalog No. 89-948-408


Add to cart

Description

Description

This gene encodes a member of the type I interferon family. Type I interferons are a group of related glycoproteins that play an important role in host defenses against viral infections. This protein is expressed in keratinocytes and the gene is found on chromosome 9, adjacent to the type I interferon cluster. [provided by RefSeq]

Sequence: VHLRRVTWQNLRHLSSMSNSFPVECLRENIAFELPQEFLQYTQPMKRDIKKAFYEMSLQAFNIFSQHTFKYWKERHLKQIQIGLDQQAEYLNQCL
Specifications

Specifications

NP_064509
Liquid
56832
IFNK (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RP11-27J8.1
IFNK
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VHLRRVTWQNLRHLSSMSNSFPVECLRENIAFELPQEFLQYTQPMKRDIKKAFYEMSLQAFNIFSQHTFKYWKERHLKQIQIGLDQQAEYLNQCL
RUO
IFNK
Wheat Germ (in vitro)
GST
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit