Learn More
Human IgA, Biotin, Isotype control, Novus Biologicals™
Human Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197181
Catalog No. NBP197181
Add to cart
Description
Description
IgA antibody specifically detects IgA in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgA
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IGHA1
IgA
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
3493
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Immunoglobulin A
Human
Ion exchange chromatography
RUO
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Human IgA (a chain specific) and anti-Human Serum. No reaction was observed against anti-Human IgG F(c) or anti-Human IgM (Fc5u).
Human