Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Human IgA, Fluorescein, Isotype control, Novus Biologicals™
Human Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197161
Catalog No. NBP197161
Description
Description
IgA antibody specifically detects IgA in Human samples. It is validated for Dot Blot.
Specifications
Specifications
IgA
Fluorescein
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Immunoglobulin A
Human
Ion exchange chromatography
RUO
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Human IgA and anti-Human Serum. No reaction was observed against anti-Human IgM Fc5u or anti-Human IgG F(c).
Human
Dot Blot
Dot Blot 1:100 - 1:2000
Conjugated
IGHA1
IgA
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
3493
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.