Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgA Antibody (BFG-267), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP142270AF700
Description
Human IgA Monoclonal specifically detects Human IgA in Human samples. It is validated for ELISA.Specifications
Human IgA | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Purified alpha-chain (Fc) of human IgA | |
0.2 mL | |
Primary | |
Human | |
Purified |
ELISA | |
BFG-267 | |
ELISA | |
Mouse | |
Ion exchange chromatography | |
RUO | |
This monoclonal antibody reacts with human IgA, alpha-chain. No cross reaction with human plasma proteins other then human IgA. No cross reaction with purified human IgE, IgG and IgM. Apparent binding constant is approximately 0.01 nM. Conjugation of the antibody with peroxidase, FITC or biotin does not impair the binding properties. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction