Learn More
Human IgA Mouse anti-Human, DyLight 350, Clone: BFG-267, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142270UV
Catalog No. NBP142270UV
Add to cart
Description
Description
Human IgA Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Human IgA
BFG-267
ELISA
Purified
purified a-chain (Fc) of human IgA. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
Ion exchange chromatography
RUO
Primary
This monoclonal antibody reacts with human IgA, alpha-chain. No cross reaction with human plasma proteins other then human IgA. No cross reaction with purified human IgE, IgG and IgM. Apparent binding constant is approximately 0.01 nM. Conjugation of the antibody with peroxidase, FITC or biotin does not impair the binding properties. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
ELISA
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
0.2 ml
Store at 4C in the dark.
Monoclonal
Human