Learn More
Abnova™ Human IGF1 (P01343, 70 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3983
Catalog No. 89-948-445
Add to cart
Description
Description
The protein encoded by this gene is similar to insulin in function and structure and is a member of a family of proteins involved in mediating growth and development. The encoded protein is processed from a precursor, bound by a specific receptor, and secreted. Defects in this gene are a cause of insulin-like growth factor I deficiency. Several transcript variants encoding different isoforms have been found for this gene
Specifications
Specifications
P01343
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
7.6kDa
Escherichia coli expression system
100 ug
RUO
IGFI
IGF1
Escherichia coli
None
Escherichia coli expression system
Lyophilized
3479
IGF1 (Human) Recombinant Protein
≥98% by SDS-PAGE and HPLC
Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at 4°C for 1 week or -20°C for longer. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
IGF1
The ED50 was determined by a cell proliferation assay using NIH3T3 cells is typically 1-7ng/mL.
Yes