Learn More
Abnova™ Human IGF2 (P01344) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4401
Catalog No. 89-948-464
Add to cart
Description
Description
This gene encodes a member of the insulin family of polypeptide growth factors that is involved in development and growth. It is an imprinted gene and is expressed only from the paternally inherited allele. It is a candidate gene for eating disorders. There is a read-through, INS-IGF2, which aligns to this gene at the 3' region and to the upstream INS gene at the 5' region. Alternatively spliced transcript variants, encoding either the same or different isoform, have been found for this gene. [provided by RefSeq]
Sequence: AYRPSETLCGGELVDTLQFVCGDRGFYFSRPASRVSRRSRGIVEECCFRSCDLALLETYCATPAKSE
Specifications
Specifications
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
7.5kDa
Escherichia coli expression system
50 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
IGF2
The activity is determined by the dose dependent proliferation of FDC-P1 cells and is typically 1.5-6ng/mL.
Yes
P01344
Lyophilized
3481
IGF2 (Human) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
AYRPSETLCGGELVDTLQFVCGDRGFYFSRPASRVSRRSRGIVEECCFRSCDLALLETYCATPAKSE
RUO
C11orf43|FLJ22066|FLJ44734|INSIGF|pp9974
IGF2
Escherichia coli
None