Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IGFBP1 Full-length ORF (AAH35263.2, 1 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name IGFBP1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 54.3kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-948-481 View Documents Abnova™
H00003484P01L
25 ug Each for $599.28
Add to cart
 
89-948-480 View Documents Abnova™
H00003484P01S
10 ug Each for $384.10
Add to cart
 
Description

Description

This gene is a member of the insulin-like growth factor binding protein (IGFBP) family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors (IGFs) I and II and circulates in the plasma. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors. [provided by RefSeq]

Sequence: MSEVPVARVWLVLLLLTVQVGVTAGAPWQCAPCSAEKLALCPPVSASCSEVTRSAGCGCCPMCALPLGAACGVATARCARGLSCRALPGEQQPLHALTRGQGACVQESDASAPHAAEAGSPESPESTEITEEELLDNFHLMAPSEEDHSILWDAISTYDGSKALHVTNIKKWKEPCRIELYRVVESLAKAQETSGEEISKFYLPNCNKNGFYHSRQCETSMDGEAGLCWCVYPWNGKRIPGSPEIRGDPNCQMYFNVQN
Specifications

Specifications

IGFBP1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
54.3kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit