Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IGFBP2 (P18065) Partial Recombinant Protein expressed in (BTI-Tn-5B1-4) Hi-5 Insect cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5988

Catalog No. 89-948-482


Add to cart

Description

Description

Sequence: EVLFRCPPCTPERLAACGPPPVAPPAAVAAVAGGARMPCAELVREPGCGCCSVCARLEGEACQVYTPRCGQGLRCYPHPGSELPLQALVMGEGTCEKRRDAEYGASPEQVADNGDDHSEGGLVENHVDSTMNMLGGGGSAGRKPLKSGMKELAVFREKVTEQHRQMGKGGKHHLGLEEPKKLRPPPARTPCQQELDQVLERISTMRLPDERGPLEHLYSLHIPNCDKHGLYNLKQCKMSLNGQRGECWCVNPNTGKLIQGAPTIRGDPECHLFYNEQQEARGVHTQRMQ
Specifications

Specifications

P18065
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
31.5kDa
Insect cell (Hi-5) expression system
20 ug
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg of protein (<1 EU/μg).
IGFBP2
Activity assay
SDS-PAGE
The optimal working dilution should be determined by the end user.
Human,Mouse ;Human,Mouse
Yes
Insect cell (Hi-5) expression system
Lyophilized
3485
IGFBP2 (Human) Recombinant Protein
0.98
EVLFRCPPCTPERLAACGPPPVAPPAAVAAVAGGARMPCAELVREPGCGCCSVCARLEGEACQVYTPRCGQGLRCYPHPGSELPLQALVMGEGTCEKRRDAEYGASPEQVADNGDDHSEGGLVENHVDSTMNMLGGGGSAGRKPLKSGMKELAVFREKVTEQHRQMGKGGKHHLGLEEPKKLRPPPARTPCQQELDQVLERISTMRLPDERGPLEHLYSLHIPNCDKHGLYNLKQCKMSLNGQRGECWCVNPNTGKLIQGAPTIRGDPECHLFYNEQQEARGVHTQRMQ
RUO
IBP2|IGF-BP53
IGFBP2
Determined by its ability to inhibit IGF-I induced proliferation of FDC-P1 cells.
Insect
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit