Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IGFBP4 (aa 22-258) His Tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$238.00 - $976.00

Specifications

Recombinant Yes
Name Human IGFBP4 (aa 22-258) His Tag
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87829 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87829
10 µg
Each for $238.00
Add to cart
 
PIRP87830 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87830
25 µg
Each for $379.00
Add to cart
 
PIRP87831 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87831
100 µg
Each for $976.00
Add to cart
 
Description

Description

This gene is a member of the insulin-like growth factor binding protein family and encodes a protein with an IGFBP domain and a thyroglobulin type-I domain. The protein binds both insulin-like growth factors I and II and circulates in the plasma in both glycosylated and non-glycosylated forms. Binding of this protein prolongs the half-life of the IGFs and alters their interaction with cell surface receptors.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative; pH 7.2
HEK293 cells
-20°C
P22692
IGFBP4
>95%
Human IGFBP4 (aa 22-258) His Tag
200 µg/mL
Liquid
AI875747; Band 3 A; BP-4; Deb2; differentially expressed in B16F10 2; HT29-IGFBP; IBP4; IBP-4; IGF-binding protein 4; IGFBP; IGFBP4; IGF-BP4; IGFBP-4; Insulin like growth factor binding protein; insulin like growth factor binding protein 4; insulin-like growth factor binding protein 4; insulin-like growth factor-binding protein 4
RUO
Human
3487
28 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit