Learn More
Abnova™ Human IGFBP6 (P24592) Partial Recombinant Protein expressed in Hi-5 (BTI-Tn-5B1-4) Insect cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6080
Catalog No. 89-948-492
Add to cart
Description
Description
Sequence: RCPGCGQGVQAGCPGGCVEEEDGGSPAEGCAEAEGCLRREGQECGVYTPNCAPGLQCHPPKDDEAPLRALLLGRGRCLPARAPAVAEENPKESKPQAGTARPQDVNRRDQQRNPGTSTTPSQPNSAGVQDTEMGPCRRHLDSVLQQLQTEVYRGAQTLYVPNCDHRGFYRKRQCRSSQGQRRGPCWCVDRMGKSLPGSPDGNGSSSCPTGSSG
Specifications
Specifications
P24592
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
22.6kDa
Insect cell (Hi-5) expression system
20 ug
Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
IGFBP6
Urease Assay
Human ;Human
Yes
Insect cell (Hi-5) expression system
Lyophilized
3489
IGFBP6 (Human) Recombinant Protein
0.95
RCPGCGQGVQAGCPGGCVEEEDGGSPAEGCAEAEGCLRREGQECGVYTPNCAPGLQCHPPKDDEAPLRALLLGRGRCLPARAPAVAEENPKESKPQAGTARPQDVNRRDQQRNPGTSTTPSQPNSAGVQDTEMGPCRRHLDSVLQQLQTEVYRGAQTLYVPNCDHRGFYRKRQCRSSQGQRRGPCWCVDRMGKSLPGSPDGNGSSSCPTGSSG
RUO
IBP6
IGFBP6
Determined by its ability to inhibit IGF-II induced proliferation of human MCF-7 cells. The expected ED50 for this effect is 0.1-0.4μg/ml.
Insect
None