Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG, Agarose Immobilized, Isotype control, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1972641MG

Catalog No. NBP1972641


Add to cart

Description

Description

IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

IgG
Agarose
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.1% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Human IgG F(c) fragment
IgG purified
RUO
Primary
Rabbit
Immunoprecipitation
Immunoprecipitation 1:10 - 1:500
Conjugated
Rabbit
IgG
1 mg
Store at 4C. Do not freeze.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit