Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG Fc Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$227.50 - $2110.00
Specifications
Antigen | IgG |
---|---|
Clone | 97924 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
IgG | |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3500 | |
Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects chimeric proteins containing a human IgG Fc region in direct ELISAs and Western blots. |
97924 | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Mouse | |
Human | |
Immunoglobulin G, ImmunoglobulinG | |
IGHG1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title