Learn More
Human IgG, Fc, Biotin, Isotype control, Novus Biologicals™
Human Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197275
Catalog No. NBP197275
Add to cart
Description
Description
IgG Fc antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
IgG Fc
LYOPH
Western Blot 1:1000, ELISA 1:5000 - 1:50000, Immunohistochemistry 1:500, Immunohistochemistry-Paraffin 1:500
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Human IgG, anti-Human IgG F(c) and anti-Human Serum. No reaction was observed against anti-Human IgG F(ab)2 or anti-Papain.
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Human
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.