Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG (H+L) Cross-Adsorbed Goat anti-Human, Alexa Fluor Plus 647, Secondary Antibody, Invitrogen™ Non-distribution product as customer accommodation.

Goat Polyclonal Antibody

$139.00 - $299.00

Specifications

Antigen Human IgG (H+L) Cross-Adsorbed
Concentration 2 mg/mL
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor Plus 647
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A48279 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
A48279
1 mg
Each for $299.00
Add to cart
 
A48279TR Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
A48279TR
100 μg
Each for $139.00
Add to cart
 
Description

Description

To minimize cross-reactivity, the goat anti-human IgG whole antibodies have been cross-adsorbed against serum proteins from bovine, mouse, and rabbit to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment to eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. This antibody binds to heavy chains on human IgG and light chains on all human immunoglobulins. This antibody does not bind non-immunoglobulin human serum proteins or serum proteins/IgG from bovine, mouse, and rabbit.

Human IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Human IgG (H+L) Cross-Adsorbed in Human samples. It is validated for Immunocytochemistry, Immunofluorescence
Specifications

Specifications

Human IgG (H+L) Cross-Adsorbed
Immunocytochemistry, Immunofluorescence
Alexa Fluor Plus 647
Goat
Human
Gamma Immunoglobins Heavy and Light chains
Polyclonal
Affinity chromatography
Secondary Antibody
2 mg/mL
Polyclonal
Liquid
RUO
Proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5
IgG
Secondary
4° C, store in dark
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit