Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IgG1 Fc, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$694.50

Specifications

Recombinant Yes
Name SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag
For Use With (Application) Control, ELISA, Western Blot
Conjugate Unlabeled
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87683 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87683
200 µg
Each for $694.50
Add to cart
 
Description

Description

Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Liquid
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
RUO
Virus
SARS-CoV-2 Spike Protein (S-RBD) (aa330-524), His Tag
Unlabeled
PBS with no preservative
E. coli
-20°Cor to +80°C if preferred
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit