Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgG2 lambda Recombinant Isotype Control (HyHEL-10)

Catalog No. MA555099
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
MA555099 100 μg
1 options

Catalog No. MA555099

Supplier: Invitrogen™ MA555099

Only null left
Add to Cart
Add to Cart

Human Recombinant Monoclonal Isotype Control

Human IgG2 lambda Isotype Control for Flow

Human IgG2, also known as human immunoglobulin G2, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the second most abundant subclass of IgG in the blood and plays a key role in defense against pathogens. IgG2 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG2 is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG2 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG2 lambda
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone HyHEL-10
Concentration 2.07 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias IGHG2; immunoglobulin heavy constant gamma 2 (G2m marker)
Host Species Human
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Target Species Not Applicable
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Isotype Control
Form Liquid
Isotype IgG2 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.