Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Invitrogen™ eBioscience™ Human IgG3 Platinum ELISA Kit Non-distribution product as customer accommodation.

For the quantitative detection of human IgG3.

Manufacturer:  Invitrogen™ BMS2094TWO

 View more versions of this product

Catalog No. 50-112-5311



Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IgG3

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Specifications

Specifications

4.7-300 ng/mL
ELISA
Adhesive Plate Covers
2 x 96 tests
0.21 ng/mL
25μL (prediluted 1:10 000)
Human
1 hr 20 min
Human
3%
HRP
HRP
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma, Serum
Plasma,10 μL|Serum,10 μL
2-8°C
1 hr 30 min
Immunoglobulin G2
5.6%
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only