Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgG4 (S228P, F234A, L235A) kappa Recombinant Isotype Control (HyHEL-10)

Catalog No. MA555108
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
MA555108 100 μg
1 options

Catalog No. MA555108

Supplier: Invitrogen™ MA555108

Only null left
Add to Cart
Add to Cart

Human Recombinant Monoclonal Isotype Control

Human IgG4 (S228P, F234A, L235A) kappa Isotype Control for Flow

Human IgG4, also known as human immunoglobulin G4, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the least abundant subclass of IgG in the blood and plays a minor role in defense against pathogens. IgG4 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG4 is also involved in other immune responses, such as immune tolerance and suppression of inflammation. Human IgG4 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG4 (S228P, F234A, L235A) kappa
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone HyHEL-10
Concentration 1.72 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias IGHG4; Immunoglobulin G; Immunoglobulin G4; immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG
Host Species Human
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Target Species Not Applicable
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Isotype Control
Form Liquid
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.