Learn More
Invitrogen™ Human IgG4 (S228P) Recombinant Isotype Control (SAA0184)
Human Recombinant Monoclonal Isotype Control
Supplier: Invitrogen™ MA555105
Description
Human IgG4 (S228P) Isotype Control for Flow
Human IgG4, also known as human immunoglobulin G4, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the least abundant subclass of IgG in the blood and plays a minor role in defense against pathogens. IgG4 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG4 is also involved in other immune responses, such as immune tolerance and suppression of inflammation. Human IgG4 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
Specifications
| Human IgG4 (S228P) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| 100 μg | |
| Not Applicable | |
| Isotype Control | |
| IgG4 |
| Flow Cytometry | |
| SAA0184 | |
| Unconjugated | |
| IGHG4; Immunoglobulin G; Immunoglobulin G4; immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG | |
| Protein A/G | |
| RUO | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.