Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human IGHG1 (P01857) Partial Recombinant Protein expressed in Hi-5 Insect cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P6023

Catalog No. 89-948-504


Add to cart

Description

Description

Sequence: LPSCKEDEYPVGSECCPKCSPGYRVKEACGELTGTVCEPCPPGTYIAHLNGLSKCLQCQMCDPAMGLRASRNCSRTENAVCGCSPGHFCIVQDGDHCAACRAYATSSPGQRVQKGGTESQDTLCQNCPPGTFSPNGTLEECQHQTKRSCDKTHTCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPGK
Specifications

Specifications

P01857
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
IGHG1-Fc (Human) Recombinant Fusion Protein
0.98
LPSCKEDEYPVGSECCPKCSPGYRVKEACGELTGTVCEPCPPGTYIAHLNGLSKCLQCQMCDPAMGLRASRNCSRTENAVCGCSPGHFCIVQDGDHCAACRAYATSSPGQRVQKGGTESQDTLCQNCPPGTFSPNGTLEECQHQTKRSCDKTHTCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPGK
RUO
IGHG1
Activity assay
SDS-PAGE
The optimal working dilution should be determined by the end user.
Human ;Human
Yes
Insect cell (Hi-5) expression system
Lyophilized
3500
Insect cell (Hi-5) expression system
20 ug
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg (<1 EU/μg).
IGHG1
Determined by its ability to neutralize 0.25ng/ml of hTNFbeta induced cytotoxicity on murine L929 cells. The expected ED50 for this effect is 1.3-1.9μg/ml of HVEM-Fc.
Insect
None
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit