Learn More
Novus Biologicals™ Human IgM Fab Mu Full length Protein
Human IgM Fab Mu Full length Protein
Manufacturer:  Novus Biologicals™ NBP1970531MG
Catalog No. NBP1970531M
Add to cart
Specifications
Specifications
1.0mg/mL
1mg
RUO - research use only
0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2
Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.