Learn More
Human IgM, HRP, Isotype control, Novus Biologicals™
Human Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197140
Catalog No. NBP197140
Add to cart
Description
Description
IgM antibody specifically detects IgM in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgM
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
Human
Ion exchange chromatography
RUO
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Human IgM, anti-Human IgG F(ab)2 and anti-Human Serum. No reaction was observed against anti-Human IgG F(c).
Human
HRP
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with No Preservative
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
IgM
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
3507
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.