Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human IgM Monoclonal Antibody (KT194)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549598
Description
This product was used in an Indirect ELISA. This product does not cross-react with human IgG1, IgG2, IgG3, IgG4, IgA and IgE.
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications
Human IgM | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P01871 | |
IGHM | |
Recombinant Human IgM. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
KT194 | |
Unconjugated | |
IGHM | |
AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
Mouse | |
Protein A | |
RUO | |
3507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction