Learn More
Human IgM Mouse anti-Human, Alexa Fluor 700, Clone: ICL-931, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142268AF700
Catalog No. NBP142268AI
Add to cart
Description
Description
Human IgM Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Immunohistochemistry (Frozen).
Specifications
Specifications
Human IgM
ICL-931
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
purified human IgM. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
Ion exchange chromatography
RUO
Primary
This monoclonal antibody reacts with human IgM, mu-chain. No cross reaction with human plasma proteins other then human IgM. No cross reaction with purified human IgA, IgE and IgG. Apparent binding constant is approximately 0.01 nM. Conjugation of the antibody with peroxidase, FITC or biotin does not impair the binding properties. Specificity of the monoclonal was determined by ELISA on purified human immunoglobulin isotypes.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
0.2 ml
Store at 4C in the dark.
Monoclonal
Human