Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgM, Texas Red, Isotype control, Novus Biologicals™
Human Antibody
Supplier: Novus Biologicals NBP197071
Description
IgM Fab Mu specifically detects IgM Fab Mu in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgM Fab Mu | |
Restore with 1.0mL of deionized water (or equivalent). | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
Multi-step | |
RUO | |
3507 | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Texas Red | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
Human | |
1 mg | |
Primary | |
No reaction was observed against anti-Human IgG F(c) or anti-Human IgA. | |
Human | |
IgM |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction