Learn More
Raybiotech Inc Human IkB-alpha ELISA Kit
SDP
Supplier:  Raybiotech Inc ELHIKBA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.65 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IkB-alpha.
Catalog No. 50-194-9388