Learn More
Raybiotech IncSupplier Diversity Partner Human IL-1 R3/IL-1 R Acp ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHIL1R31
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 18 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-1 R3.
Catalog No. 50-194-9421