Learn More
Novus Biologicals™ Human IL-10 R beta ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Supplier: Novus Biologicals™ NBP306792
This item is not returnable.
View return policy
Specifications
31.25 to 2000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human IL-10 R beta in samples. No significant cross-reactivity or interference between Human IL-10 R beta and analogues was observed. | |
3588 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |
18.75 pg/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
IL10RB | |
Cytokine Research, Immunology, Innate Immunity | |
CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 |